Sodium hydrogen sulphate
Supplier: Roth Carl
|
Danger
|
Synonyms:
Sodium bisulphate
2373.1
2373.2
ROTH2373.1EA
25.7
EUR
ROTH2373.1
ROTH2373.2
Sodium hydrogen sulphate
Sodium hydrogen sulphate
Formula:
HNaO₄S MW: 120,06 g/mol Melting Pt: 181,85 °C Density: 1,8 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003531 CAS Number: 7681-38-1 EINECS: 231-665-7 UN: 3260 ADR: 8,III Merck Index: 12,08729 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...