Marble, Supelco®

Supplier: Merck


M-Clarity™ quality level = MQ100

1.05986.1000 1.05986.5000
1.05986.1000EA 49.7 EUR
1.05986.1000 1.05986.5000
Marble, Supelco®
Marble
Formula: CaCO₃
Storage Temperature: Ambient
CAS Number: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR