Calcium carbonate

Supplier: Roth Carl

P012.1 P012.2
ROTHP012.1EA 54.1 EUR
ROTHP012.1 ROTHP012.2
Calcium carbonate
Calcium carbonate
Formula: CaCO₃
Melting Pt: 825 °C
Density: 2,71 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010906
CAS Number: 471-34-1
EINECS: 207-439-9
Merck Index: 13,01658

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR