Magnesium fluoride, Patinal®
Supplier: Merck
1.05849.0100
1.05849.1000
1.05849.0100EA
0
EUR
1.05849.0100
1.05849.1000
Magnesium fluoride, Patinal®
Magnesium fluoride
Formula:
MgF₂ MW: 62,3 g/mol Boiling Pt: 2260 °C (1013 hPa) Melting Pt: 1266 °C Density: 3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011108 CAS Number: 7783-40-6 EINECS: 231-995-1 Merck Index: 12,05701 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...