Indium(I) bromide ≥99.999% (metals basis), Puratronic®
Supplier: Thermo Scientific
|
Danger
|
42865.06
42865.14
42865.06EA
356
EUR
42865.06
42865.14
Indium(I) bromide ≥99.999% (metals basis), Puratronic®
Indium (I) bromide
Purity: 99.999%
CAS No.: 14280-53-6
Molecular Formula: InBr
Formula Weight: 194.73
Physical Form: Polycrystalline Lump
Density: 4.96
Melting Point: 220°
Boiling Point: 662°
UN No.: 3260
MDL No.: MFCD00064630
Formula:
InBr MW: 194,72 g/mol Boiling Pt: 662 °C Melting Pt: 220 °C Density: 4,96 Storage Temperature: Ambient |
MDL Number:
MFCD00064630 CAS Number: 14280-53-6 EINECS: 238-172-6 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...