Aluminium nitride, rod, round, Ø 10 mm

Supplier: Thermo Scientific

Synonyms: Aluminum nitride

42998.DR 42998.DW 42998.DO 42998.DU
42998.DREA 0 EUR
42998.DR 42998.DW 42998.DO 42998.DU
Aluminium nitride, rod, round, Ø 10 mm
Aluminium nitride

CAS No.: 24304-00-5 Physical Form: Rod Density: 3.26 Appearance: White to pale yellow Odour: Odourless Melting Point: 2200° MDL No.: MFCD00003429

Formula: AlN
MW: 40,99 g/mol
Boiling Pt: 2517 °C (1013 hPa)
Melting Pt: >2200 °C
Density: 3,26
Storage Temperature: Ambient
MDL Number: MFCD00003429
CAS Number: 24304-00-5
EINECS: 246-140-8
Merck Index: 12,00366

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR