Aluminium oxide, Patinal®

Supplier: Merck

Synonyms: Aluminium(III) oxide, Alumina C

1.01058.1000 1.01058.0100
1.01058.1000EA 0 EUR
1.01058.1000 1.01058.0100
Aluminium oxide, Patinal®
Aluminium oxide
Formula: Al₂O₃
MW: 101,96 g/mol
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3,94 g/cm³ (20 °C)
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR