Aluminium oxide, Patinal®
Supplier: Merck
Synonyms:
Aluminium(III) oxide, Alumina C
1.15345.0100
1.15345.1000
1.15345.0100EA
0
EUR
1.15345.0100
1.15345.1000
Aluminium oxide, Patinal®
Aluminium oxide
Formula:
Al₂O₃ MW: 101,96 g/mol Boiling Pt: 2980 °C (1013 hPa) Melting Pt: 2045 °C Density: 3,94 g/cm³ (20 °C) |
MDL Number:
MFCD00003424 CAS Number: 1344-28-1 EINECS: 215-691-6 REACH: 01-2119529248-35 Merck Index: 13,00355 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...