Trimethyl borate ≥99.99% (metals basis)
Supplier: Thermo Scientific
|
Danger
|
43765.22
43765.14
43765.36
43765.22EA
164
EUR
43765.22
43765.14
43765.36
Trimethyl borate ≥99.99% (metals basis)
Trimethyl borate
Purity: 99.99%
CAS No.: 121-43-7
Molecular Formula: (CH3O)3B
Formula Weight: 103.92
Flash Point: -8°(17°F)
Physical Form: Liquid
Density: 0.915
Refractive Index: 1.3610
Sensitivity: Moisture Sensitive
Appearance: Colourless
Odour: Odourless
Melting Point: -34°
Boiling Point: 67-69°
Beilstein Reference: 1697939
Merck Reference: 14,9712
UN No.: 2416
MDL No.: MFCD00008346
Formula:
C₃H₉BO₃ MW: 103,91 g/mol Boiling Pt: 67…69 °C Melting Pt: –34 °C Density: 0,915 Flash Pt: –8 °C (17 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008346 CAS Number: 121-43-7 EINECS: 204-468-9 UN: 2416 ADR: 3,II Merck Index: 14,9712 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...