Trimethyl borate for synthesis, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
8.21180.0250
8.21180.1000
8.21180.9025
8.21180.0250EA
29.3
EUR
8.21180.0250
8.21180.1000
8.21180.9025
Trimethyl borate for synthesis, Sigma-Aldrich®
Trimethyl borate
Formula:
C₃H₉BO₃ MW: 103,91 g/mol Boiling Pt: 67 °C (1013 hPa) Melting Pt: –5 °C Density: 0,915 g/cm³ (20 °C) Flash Pt: –11 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00008346 CAS Number: 121-43-7 EINECS: 204-468-9 UN: 2416 ADR: 3,II Merck Index: 12,09842 |
Specification Test Results
Solubility | (20 °C) (decomposition) |
Melting Point | -34 - -29 °C |
Molar Mass | 103.91 g/mol |
Boiling Point | 67 - 69 °C (1013 hPa) |
Vapor Pressure | 182 hPa (25 °C) |
Flash Point | -8 °C |
Refractive Index | 1.3568 (20 °C) |
Explosion Limit | 0.932 g/cm3 (20 °C) |
Density | (H2O) no data available |
Ignition Point | 305 °C |
Hazard Class | 3 |
Hazard Class Description | Flammable Liquid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...