Indium antimonide ≥99.999% (metals basis)
Supplier: Thermo Fisher Scientific
|
Warning
|
14626.04
14626.09
14626.04EA
141
EUR
14626.04
14626.09
Indium antimonide ≥99.999% (metals basis)
Indium antimonide
Purity: 99.999%
CAS No.: 1312-41-0
Molecular Formula: InSb
Formula Weight: 236.57
Physical Form: 6mm & Down Pieces
Density: 5.75 at m.p.
Melting Point: 535°
Merck Reference: 14,4948
UN No.: 1549
MDL No.: MFCD00016146
Formula:
InSb Melting Pt: 535 °C Density: 5,75 (m.p. Storage Temperature: Ambient |
MDL Number:
MFCD00016146 CAS Number: 1312-41-0 EINECS: 215-192-3 UN: 1549 ADR: 6.1,III Merck Index: 14,4948 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...