Tin(II) bromide ≥99.2%
Supplier: Thermo Fisher Scientific
Synonyms:
Tin dibromide
43430.06
43430.14
43430.06EA
34.9
EUR
43430.06
43430.14
Tin(II) bromide ≥99.2%
Tin (II) bromide
Purity: 99.2%
CAS No.: 10031-24-0
Molecular Formula: SnBr2
Formula Weight: 278.51
Physical Form: Powder
Density: 5.12
Appearance: Yellow
Melting Point: 215°
Boiling Point: 623°
Solubility: Soluble in water, with gradual decomposition. Soluble in alcohol, ether, acetone
Merck Reference: 14,8782
UN No.: 3260
MDL No.: MFCD00011239
Formula:
Br₂Sn Storage Temperature: Ambient |
MDL Number:
MFCD00011239 CAS Number: 10031-24-0 Merck Index: 14,8782 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...