Tin(II) fluoride ≥97.5%

Supplier: Thermo Scientific
Danger

11544.22 11544.36
11544.22EA 72.6 EUR
11544.22 11544.36
Tin(II) fluoride ≥97.5%
Tin (II) fluoride

Purity: 97.5% CAS No.: 7783-47-3 Molecular Formula: F2Sn Formula Weight: 156.69 Physical Form: Crystalline Density: 4.57 Appearance: White to off-white Odour: Odourless Melting Point: 219° Boiling Point: 850° Merck Reference: 14,8784 UN No.: 3260 MDL No.: MFCD00042540

Formula: SnF₂
MW: 156,71 g/mol
Boiling Pt: 850 °C
Melting Pt: 219 °C
Density: 4,57
Storage Temperature: Ambient
MDL Number: MFCD00042540
CAS Number: 7783-47-3
EINECS: 231-999-3
UN: 3260
ADR: 8,II
Merck Index: 14,8784

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR