Tin(IV) fluoride ≥99% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

13063.04 13063.09
13063.04EA 122 EUR
13063.04 13063.09
Tin(IV) fluoride ≥99% (metals basis)
Tin (IV) fluoride

Purity: 99% CAS No.: 7783-62-2 Molecular Formula: SnF4 Formula Weight: 194.68 Physical Form: -6 Mesh Crystalline Sensitivity: Moisture Sensitive Merck Reference: 14,8776 UN No.: 2923 MDL No.: MFCD00042541


Friedel-Crafts catalyst

Formula: SnF₄
MW: 194.7 g/mol
Boiling Pt: 705
Melting Pt: 705 °C (subl.)
Density: 4.78 g/cm³ (19 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042541
CAS Number: 7783-62-2
EINECS: 232-016-0
UN: 2923
ADR: 8 II
Merck Index: 14,8776

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR