Tetraiodosilane ≥99% (metals basis)

Supplier: Thermo Fisher Scientific
Danger


Packaged under argon.

87303.06 87303.14
87303.06EA 168 EUR
87303.06 87303.14
Tetraiodosilane ≥99% (metals basis)
Tetraiodosilane

Purity: 99% CAS No.: 13465-84-4 Molecular Formula: I4Si Formula Weight: 535.70 Physical Form: Crystalline, Packaged under argon Density: 4.198 Appearance: White Melting Point: 120° Boiling Point: 290° UN No.: 2923 MDL No.: MFCD00054041

Formula: I₄Si
MW: 535.7 g/mol
Boiling Pt: 290 °C
Melting Pt: 120 °C
Density: 4.198
Storage Temperature: Ambient
MDL Number: MFCD00054041
CAS Number: 13465-84-4
EINECS: 236-706-2
UN: 2923
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR