Lead(IV) oxide ≥99.995% (metals basis), Puratronic®
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Lead dioxide
10729.09
10729.18
10729.09EA
206
EUR
10729.09
10729.18
Lead(IV) oxide ≥99.995% (metals basis), Puratronic®
Lead (IV) oxide
Purity: 99.995%
CAS No.: 1309-60-0
Molecular Formula: PbO2
Formula Weight: 239.19
Physical Form: Powder
Density: 9.38
Melting Point: 290° dec.
Merck Reference: 14,5407
UN No.: 1872
MDL No.: MFCD00011165
Formula:
O₂Pb Melting Pt: 290 °C (dec.) Density: 9,38 Storage Temperature: Ambient |
MDL Number:
MFCD00011165 CAS Number: 1309-60-0 EINECS: 215-174-5 UN: 1872 ADR: 5.1,III Merck Index: 14,5407 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...