Silicon(II) oxide, Patinal®

Supplier: Merck

Synonyms: Silicon monoxide, Silicon oxide

1.10553.0100 1.10553.1000
1.10553.0100EA 0 EUR
1.10553.0100 1.10553.1000
Silicon(II) oxide, Patinal®
Silicon (II) oxide
Formula: SiO
MW: 44,08 g/mol
Boiling Pt: 1880 °C (1013 hPa)
Melting Pt: 1702 °C
Density: 2,13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00151536
CAS Number: 10097-28-6
EINECS: 233-232-8
Merck Index: 12,08640

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR