Silicon dioxide (Silica, vitreous), granules, Patinal® 1 -2.5 mm

Supplier: Merck

Synonyms: Quartz, Sand, Cristobalite

1.08044.1000 1.08044.0100
1.08044.1000EA 0 EUR
1.08044.1000 1.08044.0100
Silicon dioxide (Silica, vitreous), granules, Patinal® 1 -2.5 mm
Silicon dioxide (Silica, vitreous)
Formula: SiO₂
MW: 60,08 g/mol
Boiling Pt: 2230 °C (1013 hPa)
Melting Pt: 1710 °C
Density: 2,196 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 60676-86-0
EINECS: 262-373-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR