Methyltrichlorosilane 98+%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Trichloro(methyl)silane
127930100
127935000
ACRO127930100EA
28
EUR
ACRO127930100
ACRO127935000
Methyltrichlorosilane 98+%
Methyltrichlorosilane
Formula:
CH₃Cl₃Si MW: 149,48 g/mol Boiling Pt: 66 °C (760 mmHg) Melting Pt: –77 °C Density: 1,27 g/cm³ Flash Pt: –15 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000481 CAS Number: 75-79-6 EINECS: 200-902-6 UN: 1250 ADR: 3,II REACH: 01-2119475226-36 |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4100 to 1.4120 (20°C, 589 nm) |
GC | ≥98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...