Ethyltrimethoxysilane 97
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
Trimethoxy(ethyl)silane
B22752.14
B22752.22
B22752.14EA
29.2
EUR
B22752.14
B22752.22
Ethyltrimethoxysilane 97
Ethyltrimethoxysilane
Purity: 97%
CAS No.: 5314-55-6
Molecular Formula: (CH3O)3SiCH2CH3
Formula Weight: 150.25
Flash Point: 27°(81°F)
Refractive Index: 1.3840
Boiling Point: 123-124°
UN No.: 1993
MDL No.: MFCD00051541
Formula:
C₅H₁₄O₃Si MW: 150.25 g/mol Boiling Pt: 123…124 °C Density: 0.940 Flash Pt: 27 °C (81 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051541 CAS Number: 5314-55-6 EINECS: 226-272-9 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...