Methyltriethoxysilane 98+%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Triethoxy(methyl)silane

174622500 174620010
ACRO174622500EA 74.7 EUR
ACRO174622500 ACRO174620010
Methyltriethoxysilane 98+%
Methyltriethoxysilane
Formula: C₇H₁₈O₃Si
Boiling Pt: 141…143 °C (760 mmHg)
Melting Pt: –40 °C
Density: 0,89 g/cm³
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00009066
CAS Number: 2031-67-6
EINECS: 217-983-9
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3825 to 1.3845 (20°C, 589 nm)
GC ≥98.0 %
Hydrolyzable chloride ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR