Di-tert-butyltin dichloride ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Di-t-butyltin dichloride, Dichlorodi-tert-butylstannane
39619.02
39619.04
39619.02EA
44.2
EUR
39619.02
39619.04
Di-tert-butyltin dichloride ≥98%
Di-tert-butyltin dichloride
Purity: 98%
CAS No.: 19429-30-2
Molecular Formula: C8H18Cl2Sn
Formula Weight: 303.83
Flash Point:
Formula:
C₈H₁₈Cl₂Sn MW: 303,85 g/mol Boiling Pt: 66 °C (3 mmHg) Melting Pt: 42…43 °C Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009854 CAS Number: 19429-30-2 EINECS: 243-051-6 UN: 2928 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...