Dimethyltin dichloride 95%
Supplier: Thermo Fisher Scientific
|
Danger
|
347820050
347820250
347821000
ACRO347820050EA
71.4
EUR
ACRO347820050
ACRO347820250
ACRO347821000
Dimethyltin dichloride 95%
Dimethyltin dichloride
Formula:
C₂H₆Cl₂Sn Boiling Pt: 188…190 °C (760 mmHg) Melting Pt: 103…110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000517 CAS Number: 753-73-1 EINECS: 212-039-2 UN: 2923 ADR: 8,II |
Specification Test Results
Appearance | White to off-white Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 103 to 110 °C |
Titration Argentometric | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...