Dimethyltin dichloride

Supplier: Thermo Scientific
Danger

Synonyms: Dichlorodimethylstannane

71142.04 71142.18
71142.04EA 42.8 EUR
71142.04 71142.18
Dimethyltin dichloride
Dimethyltin dichloride

CAS No.: 753-73-1 Molecular Formula: (CH3)2SnCl2 Formula Weight: 219.69 Physical Form: Crystalline Sensitivity: Moisture Sensitive Melting Point: 103-105° Boiling Point: 185-190° Beilstein Reference: 3660508 UN No.: 3146 MDL No.: MFCD00000517

Formula: C₂H₆Cl₂Sn
MW: 219,69 g/mol
Boiling Pt: 185…190 °C
Melting Pt: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00000517
CAS Number: 753-73-1
EINECS: 212-039-2
UN: 3146
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR