Dioctyltin oxide for synthesis, Sigma-Aldrich®

Supplier: Merck

Synonyms: Di-n-octyltin oxide, Dioctyloxostannane

M-Clarity™ quality level = MQ200

8.03544.0100 8.03544.0025
8.03544.0100EA 143 EUR
8.03544.0100 8.03544.0025
Dioctyltin oxide for synthesis, Sigma-Aldrich®
Dioctyltin oxide
Formula: C₁₆H₃₄OSn
MW: 361,16 g/mol
Boiling Pt: 230 °C (1013 hPa)
Density: 1,2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013839
CAS Number: 870-08-6
EINECS: 212-791-1
UN: 3146
ADR: 6.1,III

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Molar Mass 361.15 g/mol
Bulk Density 700 kg/m3
Boiling Point 230 °C (1013 hPa)
Vapor Pressure  - 0.01 hPa (20 °C)
Flash Point 70 °C
Ignition Point 100 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR