Chlorotripropylsilane 98
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Tripropylchlorosilane
B22068.06
B22068.14
B22068.06EA
0
EUR
B22068.06
B22068.14
Chlorotripropylsilane 98
Chlorotripropylsilane
Purity: 98%
CAS No.: 995-25-5
Molecular Formula: (CH3CH2CH2)3SiCl
Formula Weight: 192.81
Flash Point: 70°(158°F)
Refractive Index: 1.4400
Odour: Acrid
Boiling Point: 199-201°
Beilstein Reference: 1737489
UN No.: 2987
MDL No.: MFCD00000508
Formula:
C₉H₂₁ClSi MW: 192.8 g/mol Boiling Pt: 199…201 °C Density: 0.881 Flash Pt: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000508 CAS Number: 995-25-5 EINECS: 213-624-5 UN: 2987 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...