Tributyltin fluoride
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Tri-n-butyltin fluoride, Fluorotributylstannane
39602.14
39602.06
39602.14EA
220
EUR
39602.14
39602.06
Tributyltin fluoride
Tributyltin fluoride
CAS No.: 1983-10-4
Molecular Formula: [CH3(CH2)3]3SnF
Formula Weight: 309.04
Physical Form: Powder
Sensitivity: Moisture Sensitive
Odour: Pungent
UN No.: 3146
MDL No.: MFCD00013547
Formula:
C₁₂H₂₇FSn MW: 309,05 g/mol Melting Pt: 248…252 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013547 CAS Number: 1983-10-4 EINECS: 217-847-9 UN: 3146 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...