Triethylsilane ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
Purity: 98+%
CAS No.: 617-86-7
Molecular Formula: C6H16Si
Formula Weight: 116.28
Flash Point: -6°(21°F)
Refractive Index: 1.4120
Melting Point: -157°
Boiling Point: 107-108°
Beilstein Reference: 1098278
UN No.: 1993
MDL No.: MFCD00009018
Formula:
C₆H₁₆Si Boiling Pt: 107…108 °C Melting Pt: –157 °C Density: 0,728 Flash Pt: –6 °C (21 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009018 CAS Number: 617-86-7 EINECS: 210-535-3 UN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...