Trimethylsilylacetylene ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Ethynyltrimethylsilane

A12856.14 A12856.06 A12856.22
A12856.14EA 226 EUR
A12856.14 A12856.06 A12856.22
Trimethylsilylacetylene ≥98%
Trimethylsilylacetylene

Purity: 98% CAS No.: 1066-54-2 Molecular Formula: C5H10Si Formula Weight: 98.22 Flash Point: -34°(-29°F) Refractive Index: 1.3900 Boiling Point: 52-53° Beilstein Reference: 906752 UN No.: 1993 MDL No.: MFCD00008569

Formula: C₅H₁₀Si
MW: 98.22 g/mol
Boiling Pt: 52…53 °C
Density: 0.710
Flash Pt: <–34 °C (–29 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00008569
CAS Number: 1066-54-2
EINECS: 213-919-9
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR