Tetrabutyltin for synthesis, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
Tetrabutylstannane, Tetra-n-butyltin
M-Clarity™ quality level = MQ200
8.08278.0025
8.08278.0100
8.08278.0025EA
25.7
EUR
8.08278.0025
8.08278.0100
Tetrabutyltin for synthesis, Sigma-Aldrich®
Tetrabutyltin
Formula:
C₁₆H₃₆Sn MW: 347,17 g/mol Boiling Pt: 145 °C (1013 hPa) Melting Pt: –97 °C Density: 1,05 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00009422 CAS Number: 1461-25-2 EINECS: 215-960-8 UN: 2788 ADR: 6.1,III |
Specification Test Results
Solubility | 0.008 g/l (20 °C) |
Melting Point | -97 °C |
Molar Mass | 347.15 g/mol |
Boiling Point | 140 °C (10 hPa) |
Vapor Pressure | 0.0014 hPa (25 °C) |
Flash Point |
|
Refractive Index | 1.4742 (20 °C, 589 nm) |
Density | 1.05 g/cm3 (20 °C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...