Tetramethylgermane ≥98%
Supplier: Thermo Fisher Scientific
|
Danger
|
A16931.06
A16931.14
A16931.06EA
184
EUR
A16931.06
A16931.14
Tetramethylgermane ≥98%
Tetramethylgermane
Purity: 98%
CAS No.: 865-52-1
Molecular Formula: Ge(CH3)4
Formula Weight: 132.73
Flash Point: -37°(-35°F)
Refractive Index: 1.3890
Melting Point: -88°
Boiling Point: 42-44°
UN No.: 1993
MDL No.: MFCD00014843
Formula:
C₄H₁₂Ge MW: 132.75 g/mol Boiling Pt: 42…44 °C Melting Pt: –88 °C Density: 0.975 Flash Pt: –37 °C (–35 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014843 CAS Number: 865-52-1 EINECS: 212-745-0 UN: 1993 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...