Phenyltrimethoxysilane ≥97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Trimethoxy(phenyl)silane
A13827.14
A13827.22
A13827.36
A13827.14EA
31.5
EUR
A13827.14
A13827.22
A13827.36
Phenyltrimethoxysilane ≥97%
Phenyltrimethoxysilane
Purity: 97%
CAS No.: 2996-92-1
Molecular Formula: C6H5Si(OCH3)3
Formula Weight: 198.30
Flash Point: 86°(186°F)
Refractive Index: 1.4735
Melting Point: -25°
Boiling Point: 210-212°
Beilstein Reference: 2937896
MDL No.: MFCD00025689
Formula:
C₉H₁₄O₃Si MW: 198,29 g/mol Boiling Pt: 210…212 °C Melting Pt: –25 °C Density: 1,070 Flash Pt: 86 °C (186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00025689 CAS Number: 2996-92-1 EINECS: 221-066-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...