Fentin chloride 95%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Chlorotriphenylstannane, Triphenyltin chloride
317291000
317290050
317295000
ACRO317291000EA
49
EUR
ACRO317291000
ACRO317290050
ACRO317295000
Fentin chloride 95%
Fentin chloride
Formula:
C₁₈H₁₅ClSn MW: 385.48 g/mol Boiling Pt: 240 °C (13.5 mmHg) Melting Pt: 103…108 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000519 CAS Number: 639-58-7 EINECS: 211-358-4 UN: 3146 ADR: 6.1 III |
Specification Test Results
Appearance | White to beige Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 103 to 108 °C |
Titration Argentometric | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...