Fentin fluoride

Supplier: Thermo Fisher Scientific
Danger

39600.03 39600.06
39600.03EA 45.7 EUR
39600.03 39600.06
Fentin fluoride
Fentin fluoride

CAS No.: 379-52-2 Molecular Formula: (C6H5)3SnF Formula Weight: 368.99 Physical Form: Powder Melting Point: 281° UN No.: 3146 MDL No.: MFCD00015356

Formula: C₁₈H₁₅FSn
MW: 369.03 g/mol
Melting Pt: >281 °C
Density: 1.5 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00015356
CAS Number: 379-52-2
EINECS: 206-833-8
UN: 3146

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR