Antimony(III) bromide 97%, pure

Supplier: Thermo Scientific
Warning

Synonyms: Antimony tribromide

318400500 318402500
ACRO318400500EA 138 EUR
ACRO318400500 ACRO318402500
Antimony(III) bromide 97%, pure
Antimony (III) bromide
Formula: Br₃Sb
MW: 361,47 g/mol
Boiling Pt: 280 °C
Melting Pt: 97 °C
Density: 2,3243 g/cm³ (50 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016317
CAS Number: 7789-61-9
EINECS: 232-179-8
UN: 1549
ADR: 6.1,III
Merck Index: 12,00747

Order Now

Specification Test Results

Appearance White to beige Crystalline powder or crystals
Titration Argentometric ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR