Antimony(III) bromide ≥99.5% (metals basis)
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Antimony tribromide
41574.09
41574.18
41574.09EA
68.1
EUR
41574.09
41574.18
Antimony(III) bromide ≥99.5% (metals basis)
Antimony (III) bromide
Purity: 99.5%
CAS No.: 7789-61-9
Molecular Formula: SbBr3
Formula Weight: 361.48
Physical Form: -8 Mesh
Density: 4.148
Refractive Index: 1.74
Sensitivity: Moisture Sensitive
Melting Point: 96.6°
Boiling Point: 280°
Merck Reference: 14,706
UN No.: 3260
MDL No.: MFCD00016317
Formula:
Br₃Sb MW: 361,47 g/mol Boiling Pt: 280 °C Melting Pt: 96,6 °C Density: 4,148 Storage Temperature: Ambient |
MDL Number:
MFCD00016317 CAS Number: 7789-61-9 EINECS: 232-179-8 UN: 3260 ADR: 8,II Merck Index: 14,706 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...