Antimony(III) fluoride ≥99%
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
Antimony trifluoride
11492.22
11492.36
11492.22EA
70.1
EUR
11492.22
11492.36
Antimony(III) fluoride ≥99%
Antimony (III) fluoride
Purity: 99+%
CAS No.: 7783-56-4
Molecular Formula: SbF3
Formula Weight: 178.75
Physical Form: Crystalline
Density: 4.379
Melting Point: 292°
Boiling Point: 376°
Merck Reference: 14,709
UN No.: 2923
MDL No.: MFCD00011218
Formula:
F₃Sb Boiling Pt: 376 °C Melting Pt: 292 °C Density: 4,379 Storage Temperature: Ambient |
MDL Number:
MFCD00011218 CAS Number: 7783-56-4 EINECS: 232-009-2 UN: 2923 ADR: 8,III Merck Index: 14,709 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...