Triethyl phosphate 99%
Supplier: Thermo Scientific
|
Warning
|
139680010
139680025
ACRO139680010EA
130
EUR
ACRO139680010
ACRO139680025
Triethyl phosphate 99%
Triethyl phosphate
Formula:
C₆H₁₅O₄P MW: 182,16 g/mol Boiling Pt: 215 °C (1013 hPa) Melting Pt: –56 °C Density: 1,07 g/cm³ Flash Pt: 111 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009077 CAS Number: 78-40-0 EINECS: 201-114-5 Merck Index: 12,09806 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4050 to 1.4070 (20°C, 589 nm) |
Water | ≤0.2 % |
GC | ≥98.5 % |
Color scale | ≤20 APHA |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...