Trimethyl phosphate 99%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
Trimethylphosphate
157970500
157975000
157970025
ACRO157970500EA
25.4
EUR
ACRO157970500
ACRO157975000
ACRO157970025
Trimethyl phosphate 99%
Trimethyl phosphate
Formula:
C₃H₉O₄P MW: 140,08 g/mol Boiling Pt: 197 °C (760 mmHg) Melting Pt: –46 °C Density: 1,197 g/cm³ Flash Pt: 107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008348 CAS Number: 512-56-1 EINECS: 208-144-8 |
Specification Test Results
Appearance | Clear liquid |
Infrared spectrum | Conforms |
Refractive index | 1.3930 to 1.3970 (20°C, 589 nm) |
Water | ≤0.2 % |
GC | ≥98.5 % |
Color scale | ≤30 APHA |
Residue after evaporation | ≤0.001 % |
Acidity | ≤0.0005 meq/g |
Alkalinity | ≤0.0002 meq/g |
Aluminium (Al) | ≤0.5 ppm |
Barium (Ba) | ≤0.1 ppm |
Boron (B) | ≤0.02 ppm |
Cadmium (Cd) | ≤0.05 ppm |
Calcium (Ca) | ≤0.5 ppm |
Chromium (Cr) | ≤0.02 ppm |
Cobalt (Co) | ≤0.02 ppm |
Copper (Cu) | ≤0.02 ppm |
Iron (Fe) | ≤0.1 ppm |
Lead (Pb) | ≤0.1 ppm |
Magnesium (Mg) | ≤0.1 ppm |
Manganese (Mn) | ≤0.02 ppm |
Nickel (Ni) | ≤0.02 ppm |
Tin (Sn) | ≤0.1 ppm |
Zinc (Zn) | ≤0.1 ppm |
Carbonyl compounds | ≤0.01 % (as butyraldehyde) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...