Trimethyl phosphate ≥98%

Supplier: Thermo Scientific
Danger

Synonyms: Trimethylphosphate

A10595.36 A10595.0E
A10595.36EA 72.3 EUR
A10595.36 A10595.0E
Trimethyl phosphate ≥98%
Trimethyl phosphate

Purity: 98+% CAS No.: 512-56-1 Molecular Formula: C3H9O4P Formula Weight: 140.08 Flash Point: 150°(302°F) Refractive Index: 1.3970 Odour: Pleasant Melting Point: -46° Boiling Point: 196-198° Beilstein Reference: 1071731 MDL No.: MFCD00008348

Formula: C₃H₉O₄P
MW: 140,08 g/mol
Boiling Pt: 196…198 °C
Melting Pt: –46 °C
Density: 1,213
Flash Pt: >150 °C (302 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008348
CAS Number: 512-56-1
EINECS: 208-144-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR