Triphenyl phosphite 99%

Supplier: Thermo Scientific
Warning

140440010 140440025 140440100
ACRO140440010EA 99.7 EUR
ACRO140440010 ACRO140440025 ACRO140440100
Triphenyl phosphite 99%
Triphenyl phosphite
Formula: C₁₈H₁₅O₃P
Boiling Pt: 360 °C (760 mmHg)
Melting Pt: 22…24 °C
Density: 1,184 g/cm³
Flash Pt: 146 °C
Storage Temperature: Ambient
MDL Number: MFCD00003032
CAS Number: 101-02-0
EINECS: 202-908-4
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5890 to 1.5920 (20°C, 589 nm)
GC ≥98.5 %
Acid value ≤1.5 mg KOH/g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR