Trioctylphosphine tech. 90%, Technical Grade
Supplier: Thermo Fisher Scientific
|
Danger
|
Synonyms:
TOP, Tri-n-octylphosphine
L15744.AC
L15744.AE
L15744.AP
L15744.ACEA
65.1
EUR
L15744.AC
L15744.AE
L15744.AP
Trioctylphosphine tech. 90%, Technical Grade
Trioctylphosphine
Purity: 90%
CAS No.: 4731-53-7
Molecular Formula: C24H51P
Formula Weight: 370.65
Flash Point: 147°(296°F)
Refractive Index: 1.4680
Odour: Garlic-like
Boiling Point: 240°/10mm
Beilstein Reference: 1776995
UN No.: 1760
MDL No.: MFCD00015298
Formula:
C₂₄H₅₁P MW: 370.64 g/mol Boiling Pt: 240 °C (10 mmHg) Melting Pt: 48 Density: 0.831 Flash Pt: 147 °C (296 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00015298 CAS Number: 4731-53-7 EINECS: 225-234-2 UN: 1760 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...