Cetrimonium bromide

Supplier: Roth Carl
Danger

9161.1 9161.2
ROTH9161.1EA 36.2 EUR
ROTH9161.1 ROTH9161.2
Cetrimonium bromide
Cetrimonium bromide
Formula: CH₃(CH₂)₁₅N(Br)(CH₃)₃
MW: 364,45 g/mol
Melting Pt: 237…243 °C
Flash Pt: 244 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00011772
CAS Number: 57-09-0
EINECS: 200-311-3
UN: 3077
ADR: 9,III
Merck Index: 13,02034

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR