Chlormequat chloride 98%

Supplier: Thermo Fisher Scientific
Danger

215420250 215421000
ACRO215420250EA 46.2 EUR
ACRO215420250 ACRO215421000
Chlormequat chloride 98%
Chlormequat chloride
Formula: ClCH₂CH₂N(CH₃)₃Cl
Melting Pt: 239…243 °C
Storage Temperature: Ambient
MDL Number: MFCD00011869
CAS Number: 999-81-5
EINECS: 213-666-4
UN: 2811
ADR: 6.1,III
Merck Index: 13,02120

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder or crystals
Infrared spectrum Conforms
Water ≤1 % (K.F.)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR