Acetylthiocholine iodide 98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
(2-Mercaptoethyl)trimethylammonium iodide acetate, 2-(Acetylthio)-N,N,N-trimethylethan-1-aminium iodide, S-Acetylthiocholine iodide
102710050
102710250
ACRO102710050EA
150
EUR
ACRO102710050
ACRO102710250
Acetylthiocholine iodide 98%
Acetylthiocholine iodide
Formula:
H₃CC(S)O(CH₂)₂N(CH₃)₃I MW: 289,18 g/mol Melting Pt: 205…210 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00011819 CAS Number: 1866-15-5 EINECS: 217-474-1 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Titration Argentometric | ≥97.5 % |
Titration with HClO4 | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...