Ammonium chloride
Supplier: Roth Carl
|
Warning
|
Synonyms:
Ammonium muriate, Salmiac
P726.1
P726.2
P726.5
ROTHP726.1EA
31.7
EUR
ROTHP726.1
ROTHP726.2
ROTHP726.5
Ammonium chloride
Ammonium chloride
Formula:
NH₄Cl Melting Pt: 335 °C Density: 1,5256 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011420 CAS Number: 12125-02-9 EINECS: 235-186-4 Merck Index: 13,00510 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...