Ammonium hydrogen carbonate
Supplier: Roth Carl
|
Warning
|
Synonyms:
Ammonium bicarbonate
T871.1
T871.2
T871.5
ROTHT871.1EA
38.6
EUR
ROTHT871.1
ROTHT871.2
ROTHT871.5
Ammonium hydrogen carbonate
Ammonium hydrogen carbonate
Formula:
NH₄HCO₃ MW: 79.06 g/mol Melting Pt: 35 Density: 1.57 g/cm³ (20 °C) |
MDL Number:
MFCD00012138 CAS Number: 1066-33-7 EINECS: 213-911-5 Merck Index: 13,00497 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...