Tetramethylthiuram monosulphide ≥97%

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Bis(dimethylthiocarbamyl) sulphide

A17259.14 A17259.22
A17259.14EA 26.8 EUR
A17259.14 A17259.22
Tetramethylthiuram monosulphide ≥97%
Tetramethylthiuram monosulphide

Purity: 97% CAS No.: 97-74-5 Molecular Formula: C6H12N2S3 Formula Weight: 208.37 Melting Point: 106-110° Beilstein Reference: 1775650

Formula: C₆H₁₂N₂S₃
MW: 208.37 g/mol
Melting Pt: 106…110 °C
Density: 1.37
Storage Temperature: Ambient
MDL Number: MFCD00014870
CAS Number: 97-74-5
EINECS: 202-605-7
UN: 3077

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR