Dipentyl sulphide ≥97%
Supplier: Thermo Fisher Scientific
|
Warning
|
Purity: 97%
CAS No.: 872-10-6
Molecular Formula: C10H22S
Formula Weight: 174.35
Flash Point: 85°(185°F)
Refractive Index: 1.4560
Odour: Stench
Melting Point: -51°
Boiling Point: 228-230°
Beilstein Reference: 1698031
MDL No.: MFCD00027275
Formula:
C₁₀H₂₂S Boiling Pt: 228…230 °C Melting Pt: –51 °C Density: 0,840 Flash Pt: 85 °C (185 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00027275 CAS Number: 872-10-6 EINECS: 212-820-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...