Diphenyl disulphide ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
Synonyms:
1,2-Diphenyldisulphane, Phenyl disulphide
A12586.18
A12586.30
A12586.18EA
46.8
EUR
A12586.18
A12586.30
Diphenyl disulphide ≥98%
Diphenyl disulphide
Purity: 98%
CAS No.: 882-33-7
Molecular Formula: C12H10S2
Formula Weight: 218.34
Odour: Unpleasant
Melting Point: 58-62°
Boiling Point: 310°
Beilstein Reference: 639794
MDL No.: MFCD00003065
Formula:
C₁₂H₁₀S₂ MW: 218.34 g/mol Boiling Pt: 310 °C Melting Pt: 58…62 °C Density: 1.155 g/cm³ (64.9 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003065 CAS Number: 882-33-7 EINECS: 212-926-4 UN: 3335 ADR: 9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...